Search
Search

Adcy3| Gene Symbol: Adcy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130779.2 | NP_570135.2 | 2 | 791 | ||
| XM_006239847.2 | XP_006239909.1 | 1 | 527 | |||
| XM_006239848.2 | XP_006239910.1 | 1 | 530 | |||
| XM_006239849.2 | XP_006239911.1 | 1 | 524 | |||
| XM_006239850.2 | XP_006239912.1 | 1 | 533 | |||
| XM_008764532.1 | XP_008762754.1 | 2 | 647 | |||
| GenBank mRNA | M55075.1 | - | 2 | 716 | ||