Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031050.1 | NP_112312.1 | 3 | 951 | ||
GenBank mRNA | BC061878.1 | - | 2 | 945 | 5599252 | |
FQ221670.1 | - | 3 | 1018 | |||
FQ229097.1 | - | 3 | 1014 | |||
FQ230118.1 | - | 2 | 780 | |||
X84039.1 | - | 3 | 951 |