Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031132.3 | NP_112394.3 | 4 | 796 | ||
XM_008766690.1 | XP_008764912.1 | 5 | 1352 | |||
GenBank mRNA | GU067683.1 | - | 4 | 545 | ||
L09653.1 | - | 4 | 603 | |||
S67770.1 | - | 4 | 796 |