Search
Search

Kcnj4| Gene Symbol: Kcnj4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053870.2 | NP_446322.2 | 2 | 1328 | ||
| XM_006241963.2 | XP_006242025.1 | 1 | 773 | |||
| XM_006241964.2 | XP_006242026.1 | 2 | 2053 | |||
| GenBank mRNA | U27582.1 | - | 2 | 1093 | ||
| X83580.1 | - | 1 | 730 | |||
| X87635.1 | - | 2 | 1328 | |||