Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139255.3 | NP_640348.2 | 11 | 1530 | ||
GenBank mRNA | AY095481.1 | - | 5 | 562 | ||
BC086408.1 | - | 6 | 663 | |||
BC097934.1 | - | 7 | 836 | |||
BC104715.1 | - | 6 | 601 | |||
BC126078.1 | - | 7 | 734 | |||
BC161835.1 | - | 7 | 692 |