Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127201.1 | NP_001120673.1 | 4 | 517 | ||
XM_006227721.2 | XP_006227783.1 | 4 | 550 | |||
XM_006227722.2 | XP_006227784.1 | 4 | 474 | |||
XM_008758627.1 | XP_008756849.1 | 5 | 517 | |||
GenBank mRNA | CF109177.1 | - | 3 | 209 | ||
FQ213058.1 | - | 3 | 169 | |||
FQ220549.1 | - | 3 | 169 | |||
FQ234587.1 | - | 4 | 535 |