Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BMP2 SIRNA Assays ›
Gene Symbol:
Bmp2
Gene Name:
bone morphogenetic protein 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 126335963 - 126345881 on Build Rnor_6.0
UniGene ID:
Rn.90931
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS334240
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Bmp2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017178.1
NP_058874.1
3
629
XM_008762246.1
XP_008760468.1
2
1898
GenBank mRNA
L20678.1
-
3
629
Z25868.1
-
3
629
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007553, AK161862, AK133923, BC100344, AK132433.
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
activation of MAPK activity
ossification
osteoblast differentiation
branching involved in ureteric bud morphogenesis
response to hypoxia
in utero embryonic development
epithelial to mesenchymal transition
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
chondrocyte differentiation
BMP signaling pathway involved in heart induction
atrioventricular valve morphogenesis
endocardial cushion morphogenesis
negative regulation of Wnt signaling pathway involved in heart development
proteoglycan metabolic process
regulation of transcription, DNA-templated
protein phosphorylation
inflammatory response
Notch signaling pathway
heart development
negative regulation of cell proliferation
response to mechanical stimulus
organ morphogenesis
positive regulation of gene expression
negative regulation of gene expression
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of steroid biosynthetic process
positive regulation of phosphatase activity
telencephalon development
telencephalon regionalization
positive regulation of Wnt signaling pathway
bone mineralization
positive regulation of cell migration
positive regulation of bone mineralization
BMP signaling pathway
protein destabilization
negative regulation of aldosterone biosynthetic process
response to retinoic acid
positive regulation of osteoblast proliferation
cardiocyte differentiation
embryonic heart tube anterior/posterior pattern specification
bone mineralization involved in bone maturation
growth
odontogenesis of dentin-containing tooth
positive regulation of odontogenesis
regulation of odontogenesis of dentin-containing tooth
ovulation cycle
regulation of apoptotic process
positive regulation of apoptotic process
regulation of MAPK cascade
positive regulation of MAPK cascade
negative regulation of insulin-like growth factor receptor signaling pathway
cell fate commitment
positive regulation of cell differentiation
positive regulation of fat cell differentiation
positive regulation of neuron differentiation
positive regulation of osteoblast differentiation
positive regulation of ossification
negative regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of astrocyte differentiation
mesenchymal cell differentiation
inner ear development
positive regulation of neurogenesis
negative regulation of calcium-independent cell-cell adhesion
cartilage development
cardiac muscle cell differentiation
cardiac muscle tissue morphogenesis
oxidation-reduction process
pericardium development
corticotropin hormone secreting cell differentiation
thyroid-stimulating hormone-secreting cell differentiation
cardiac epithelial to mesenchymal transition
pathway-restricted SMAD protein phosphorylation
SMAD protein signal transduction
mesenchyme development
positive regulation of Wnt signaling pathway by BMP signaling pathway
positive regulation of cartilage development
positive regulation of ERK1 and ERK2 cascade
cellular response to mechanical stimulus
cellular response to growth factor stimulus
cellular response to organic cyclic compound
cellular response to BMP stimulus
mesenchymal cell proliferation involved in ureteric bud development
negative regulation of canonical Wnt signaling pathway
positive regulation of p38MAPK cascade
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
negative regulation of cortisol biosynthetic process
negative regulation of cardiac muscle cell differentiation
retinol dehydrogenase activity
receptor binding
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
phosphatase activator activity
protein domain specific binding
protein homodimerization activity
SMAD binding
protein heterodimerization activity
BMP receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0