siRNA Details
Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_217313.8 | XP_217313.6 | 1 | 1873 | ||
GenBank mRNA | FQ231323.1 | - | 1 | 160 |

Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_217313.8 | XP_217313.6 | 1 | 1873 | ||
GenBank mRNA | FQ231323.1 | - | 1 | 160 |