Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230595.2 | XP_006230657.1 | 2 | 1628 | ||
XM_006230596.2 | XP_006230658.1 | 3 | 992 | |||
XM_006230597.2 | XP_006230659.1 | 2 | 880 |
Gene Symbol: Cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127541.1 | NP_001121013.1 | 255 | |||
XM_006230559.2 | XP_006230621.1 | 2 | 1628 | |||
XM_006230561.2 | XP_006230623.1 | 2 | 880 | |||
GenBank mRNA | BC158864.1 | - | 255 | 7936496 |