Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DDX3Y SIRNA Assays ›
Gene Symbol:
Ddx3y
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box helicase 3, Y-linked
Gene Aliases:
RGD1309586
Chromosome Location:
Chr.13: 105408179 - 105411348 on Build Rnor_6.0
UniGene ID:
Rn.218622
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS338563
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ddx3y
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001108858.1
NP_001102328.1
1
1006
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
chromosome segregation
metabolic process
extrinsic apoptotic signaling pathway via death domain receptors
response to virus
RNA secondary structure unwinding
positive regulation of gene expression
negative regulation of translation
positive regulation of cell growth
negative regulation of cell growth
negative regulation of protein complex assembly
DNA duplex unwinding
stress granule assembly
intracellular signal transduction
mature ribosome assembly
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of viral genome replication
innate immune response
positive regulation of translation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of translational initiation
cellular response to arsenic-containing substance
cellular response to osmotic stress
intrinsic apoptotic signaling pathway
positive regulation of G1/S transition of mitotic cell cycle
negative regulation of intrinsic apoptotic signaling pathway
nucleic acid binding
DNA binding
RNA binding
GTPase activity
ATP-dependent DNA helicase activity
ATP-dependent RNA helicase activity
helicase activity
ATP binding
transcription factor binding
poly(A) binding
eukaryotic initiation factor 4E binding
ATPase activity
nucleoside-triphosphatase activity
translation initiation factor binding
RNA stem-loop binding
ribosomal small subunit binding
CTPase activity
poly(A) RNA binding
mRNA 5'-UTR binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-cdcg2:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0