Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271381.1 | NP_001258310.1 | 8 | 632 | ||
XM_008772413.1 | XP_008770635.1 | 7 | 797 | |||
GenBank mRNA | BC168970.1 | - | 7 | 994 |