Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRKCQ SIRNA Assays ›
Gene Symbol:
Prkcq
Gene Name:
protein kinase C, theta
Gene Aliases:
Pkcq
Chromosome Location:
Chr.17: 70971915 - 71105286 on Build Rnor_6.0
UniGene ID:
Rn.8227
Species:
Rat
siRNA ID
RSS340431
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Prkcq
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276721.1
NP_001263650.1
18
2221
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
immune system process
regulation of transcription, DNA-templated
protein phosphorylation
membrane protein ectodomain proteolysis
inflammatory response
aging
response to heat
response to glucose
regulation of G2/M transition of mitotic cell cycle
response to organic cyclic compound
regulation of vasoconstriction
neuron differentiation
positive regulation of interleukin-17 production
positive regulation of interleukin-4 production
response to insulin
intracellular signal transduction
positive regulation of T cell proliferation
tissue regeneration
positive regulation of NF-kappaB import into nucleus
positive regulation of interleukin-2 biosynthetic process
respiratory burst
negative regulation of insulin receptor signaling pathway
positive regulation of protein secretion
positive regulation of T cell activation
positive regulation of NF-kappaB transcription factor activity
positive regulation of filopodium assembly
positive regulation of stress fiber assembly
negative regulation of T cell apoptotic process
regulation of platelet aggregation
positive regulation of T-helper 17 type immune response
positive regulation of T-helper 2 cell activation
protein serine/threonine kinase activity
protein kinase C activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-4vcgr:80/10.239.160.220:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0