Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106059.1 | NP_001099529.1 | 2 | 421 | ||
GenBank mRNA | BC168852.1 | - | 2 | 404 | 7443262 |