Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191719.1 | NP_001178648.1 | 8 | 2196 | ||
XM_006256359.2 | XP_006256421.1 | 8 | 2247 | |||
XM_006256360.2 | XP_006256422.1 | 7 | 2043 | |||
XM_006256361.2 | XP_006256423.1 | 8 | 2270 | |||
XM_006256362.2 | XP_006256424.1 | 8 | 2162 | |||
XM_006256363.1 | XP_006256425.1 | 4 | 1604 | |||
XM_006256364.1 | XP_006256426.1 | 8 | 2156 | |||
XM_006256365.1 | XP_006256427.1 | 3 | 1462 |