Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gchfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133595.2 | NP_598279.1 | 3 | 273 | ||
GenBank mRNA | BC086944.1 | - | 3 | 254 | 7325198 | |
U53710.1 | - | 3 | 273 | |||
U85512.1 | - | 3 | 300 |