Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134365.2 | NP_599192.1 | 6 | 775 | ||
GenBank mRNA | BC063808.1 | - | 6 | 716 | 6920021 | |
CK845764.1 | - | 443 | ||||
FQ209675.1 | - | 6 | 674 | |||
FQ212544.1 | - | 6 | 711 | |||
FQ214336.1 | - | 6 | 703 | |||
FQ214353.1 | - | 6 | 737 | |||
FQ214371.1 | - | 6 | 711 | |||
FQ214449.1 | - | 6 | 674 | |||
FQ214672.1 | - | 6 | 710 | |||
FQ214784.1 | - | 6 | 709 | |||
FQ215244.1 | - | 6 | 674 | |||
FQ215436.1 | - | 6 | 674 | |||
FQ215480.1 | - | 6 | 674 | |||
FQ215823.1 | - | 6 | 727 | |||
FQ216247.1 | - | 6 | 660 | |||
FQ216597.1 | - | 6 | 666 | |||
FQ216773.1 | - | 6 | 674 | |||
FQ216829.1 | - | 6 | 674 | |||
FQ216922.1 | - | 6 | 674 | |||
FQ218701.1 | - | 6 | 710 | |||
FQ219899.1 | - | 6 | 703 | |||
FQ223652.1 | - | 6 | 674 | |||
FQ223683.1 | - | 6 | 700 | |||
FQ229261.1 | - | 6 | 734 | |||
FQ229976.1 | - | 6 | 711 | |||
FQ232356.1 | - | 6 | 711 | |||
M35052.1 | - | 6 | 694 |
Gene Symbol: LOC100911417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749372.2 | XP_003749420.1 | 770 |