Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105957.1 | NP_001099427.1 | 18 | 2465 | ||
XM_006249982.2 | XP_006250044.1 | 17 | 2579 | |||
XM_008769609.1 | XP_008767831.1 | 17 | 2228 | |||
XM_008769610.1 | XP_008767832.1 | 16 | 2117 |