Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106135.2 | NP_001099605.2 | 2 | 192 | ||
XM_006254716.2 | XP_006254778.1 | 2 | 218 | |||
GenBank mRNA | BC166853.1 | - | 2 | 167 | 9092757 | |
CO565239.1 | - | 2 | 192 |