Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cercam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011962.1 | NP_001011962.1 | 13, 14 | 1500 | ||
XM_006233768.2 | XP_006233830.1 | 10, 11 | 2168 | |||
XM_006233769.2 | XP_006233831.1 | 11, 12 | 1560 | |||
XM_006233770.2 | XP_006233832.1 | 13, 14 | 2540 | |||
XM_006233771.2 | XP_006233833.1 | 9, 10 | 2034 | |||
GenBank mRNA | BC085782.1 | - | 13, 14 | 1500 | 7107449 |