Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008513.1 | NP_001008513.1 | 2 | 203 | ||
GenBank mRNA | BC086571.1 | - | 2 | 203 | 7304213 | |
FQ218040.1 | - | 2 | 227 | |||
FQ220091.1 | - | 2 | 227 | |||
FQ223815.1 | - | 2 | 229 | |||
FQ225478.1 | - | 2 | 227 | |||
FQ226079.1 | - | 2 | 225 | |||
FQ233622.1 | - | 2 | 227 |