Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nif3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024763.1 | NP_001019934.1 | 2 | 565 | ||
XM_006244958.2 | XP_006245020.1 | 4 | 706 | |||
XM_006244960.2 | XP_006245022.1 | 4 | 677 | |||
XM_006244961.2 | XP_006245023.1 | 4 | 702 | |||
XM_006244962.2 | XP_006245024.1 | 3 | 575 | |||
XM_006244963.2 | XP_006245025.1 | 4 | 709 | |||
XM_008767082.1 | XP_008765304.1 | 4 | 710 | |||
GenBank mRNA | BC097437.1 | - | 2 | 565 | 7455562 | |
BC097992.1 | - | 4 | 638 | 7446502 |