siRNA Details
Gene Symbol: LOC102548927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773285.1 | XP_008771507.1 | 1 | 490 |

Gene Symbol: LOC102548927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773285.1 | XP_008771507.1 | 1 | 490 |