Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025670.1 | NP_001020841.1 | 7, 8 | 1145 | ||
XM_008770019.1 | XP_008768241.1 | 8, 9 | 1142 | |||
XM_008770020.1 | XP_008768242.1 | 8, 9 | 1571 | |||
XM_008770021.1 | XP_008768243.1 | 8, 9 | 1270 | |||
GenBank mRNA | BC097362.1 | - | 7, 8 | 1145 | 7442656 |