Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012148.1 | NP_001012148.1 | 12 | 1392 | ||
XM_006249461.1 | XP_006249523.1 | 12 | 1932 | |||
XM_006249462.2 | XP_006249524.1 | 12 | 1276 | |||
GenBank mRNA | BC085774.1 | - | 12 | 1392 | 7096477 |