Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Natd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170541.1 | NP_001164012.1 | 2 | 170 | ||
GenBank mRNA | CB546029.1 | - | 2 | 144 | ||
CK480689.1 | - | 2 | 170 | |||
FQ231549.1 | - | 2 | 212 |
Gene Symbol: LOC100911674 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750836.3 | XP_003750884.1 | 2 | 900 |