Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tifab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025029.1 | NP_001020200.1 | 3 | 672 | ||
XM_006253578.2 | XP_006253640.1 | 3 | 1084 | |||
XM_006253579.2 | XP_006253641.1 | 3 | 973 | |||
XM_008771443.1 | XP_008769665.1 | 2 | 2230 | |||
GenBank mRNA | BC091188.1 | - | 3 | 672 | 7367314 |