Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134781.1 | NP_001128253.1 | 3 | 718 | ||
XM_006236049.2 | XP_006236111.1 | 3 | 970 | |||
XM_006236050.2 | XP_006236112.1 | 4 | 1824 | |||
XM_006236051.2 | XP_006236113.1 | 3 | 420 | |||
XM_008762709.1 | XP_008760931.1 | 3 | 1942 | |||
XM_008762710.1 | XP_008760932.1 | 4 | 1336 | |||
XM_008762711.1 | XP_008760933.1 | 3 | 1942 | |||
GenBank mRNA | BC158823.1 | - | 3 | 719 |