Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109033.2 | NP_001102503.1 | 2 | 818 | ||
XM_006246776.2 | XP_006246838.1 | 3 | 1717 | |||
XM_008767852.1 | XP_008766074.1 | 2 | 2140 | |||
GenBank mRNA | BC158839.1 | - | 2 | 818 | 7387741 |