siRNA Details
Gene Symbol: Ccdc54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_573274.4 | XP_573274.1 | 1 | 855 |

Gene Symbol: Ccdc54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_573274.4 | XP_573274.1 | 1 | 855 |