siRNA Details
Gene Symbol: Shisa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191936.1 | NP_001178865.1 | 2 | 605 |

Gene Symbol: Shisa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191936.1 | NP_001178865.1 | 2 | 605 |