Search
Search

Ccdc17| Gene Symbol: Ccdc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014067.2 | NP_001014089.2 | 9 | 1130 | ||
| XM_006238702.2 | XP_006238764.1 | 8 | 1360 | |||
| XM_006238703.2 | XP_006238765.1 | 8 | 1360 | |||
| XM_006238704.2 | XP_006238766.1 | 9 | 1288 | |||
| XM_006238705.2 | XP_006238767.1 | 8 | 1360 | |||
| XM_006238707.2 | XP_006238769.1 | 9 | 1288 | |||
| GenBank mRNA | BC087059.1 | - | 7 | 1374 | ||
| CO398762.1 | - | 5 | 412 | |||