siRNA Details
Gene Symbol: Hbe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024805.1 | NP_001019976.1 | 2, 3 | 312 |

Gene Symbol: Hbe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024805.1 | NP_001019976.1 | 2, 3 | 312 |