Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pramel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109368.1 | NP_001102838.1 | 3 | 1202 |
Gene Symbol: LOC100910698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752110.3 | XP_003752158.1 | 8 | 1713 | ||
XM_008773459.1 | XP_008771681.1 | 9 | 1641 |