Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023971.1 | NP_076461.1 | 13 | 1617 | ||
XM_006257235.2 | XP_006257297.1 | 12 | 1588 | |||
XM_008773432.1 | XP_008771654.1 | 13 | 1744 | |||
XM_008773433.1 | XP_008771655.1 | 13 | 1500 | |||
XM_008773434.1 | XP_008771656.1 | 13 | 1715 | |||
XM_008773435.1 | XP_008771657.1 | 12 | 1588 | |||
XM_008773436.1 | XP_008771658.1 | 11 | 3053 | |||
GenBank mRNA | AF195787.1 | - | 13 | 1617 | ||
AF195788.1 | - | 12 | 1550 |