Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750162.3 | XP_003750210.1 | 9 | 1008 | ||
XM_008764676.1 | XP_008762898.1 | 9 | 1008 | |||
XM_008764677.1 | XP_008762899.1 | 10 | 1439 | |||
GenBank mRNA | FQ220776.1 | - | 4 | 325 |