Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100158.2 | NP_001093628.1 | 7, 8 | 793 | ||
GenBank mRNA | BC105875.1 | - | 7, 8 | 738 | 7130292 | |
FQ234161.1 | - | 6, 7 | 514 |