Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099516.1 | NP_001092986.1 | 1778 |
Gene Symbol: LOC100911147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748752.1 | XP_003748800.1 | 6 | 1769 | ||
XM_006228209.1 | XP_006228271.1 | 6 | 1772 | |||
XM_006228210.1 | XP_006228272.1 | 6 | 1775 | |||
XM_006228211.1 | XP_006228273.1 | 6 | 1772 |
Set Membership: |
G-protein coupled receptor |