Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP53 SIRNA Assays ›
Gene Symbol:
TP53
Gene Name:
tumor protein p53
Gene Aliases:
BCC7, LFS1, P53, TRP53
Chromosome Location:
Chr.17: 7668402 - 7687550 on Build GRCh38
UniGene ID:
Hs.437460
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
VHS40366
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TP53
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000546.5
NP_000537.3
7
904
NM_001126112.2
NP_001119584.1
7
901
NM_001126113.2
NP_001119585.1
7
904
NM_001126114.2
NP_001119586.1
7
904
NM_001126115.1
NP_001119587.1
3
584
NM_001126116.1
NP_001119588.1
3
584
NM_001126117.1
NP_001119589.1
3
584
NM_001126118.1
NP_001119590.1
6
1021
NM_001276695.1
NP_001263624.1
7
904
NM_001276696.1
NP_001263625.1
7
904
NM_001276697.1
NP_001263626.1
3
584
NM_001276698.1
NP_001263627.1
3
584
NM_001276699.1
NP_001263628.1
3
584
NM_001276760.1
NP_001263689.1
7
904
NM_001276761.1
NP_001263690.1
7
901
GenBank mRNA
AB082923.1
-
7
765
AF307851.1
-
7
837
AK225838.1
-
7
842
AK297462.1
-
8
769
AK297927.1
-
8
814
AK312568.1
-
7
846
AM076970.1
-
6
669
AM076971.1
-
6
702
AM076972.1
-
6
702
AY429684.1
-
6
702
BC003596.1
-
7
822
3544714
BT019622.1
-
6
702
DQ186648.1
-
7
834
DQ186649.1
-
7
834
DQ186650.1
-
3
580
DQ186651.1
-
3
580
DQ186652.1
-
3
580
DQ191317.1
-
7
891
DQ263704.1
-
6
702
DQ286964.1
-
7
836
DQ648884.1
-
6
502
EF101867.1
-
6
669
EF101868.1
-
6
702
EF101869.1
-
6
702
FJ207420.1
-
7
837
JN214348.1
-
6
747
JN601513.1
-
6
702
JN601514.1
-
6
702
JN601515.1
-
6
702
JN601516.1
-
7
676
JN900492.1
-
6
1010
JQ026207.1
-
6
692
JQ026208.1
-
6
697
JQ026209.1
-
6
707
JQ026210.1
-
6
702
JQ694049.1
-
6
725
JQ694050.1
-
6
725
JQ694051.1
-
6
725
KC820708.1
-
3
197
KC820709.1
-
3
197
KC820710.1
-
3
197
KC820711.1
-
3
197
KC820712.1
-
3
197
KC820713.1
-
3
197
KC820714.1
-
3
197
KC820715.1
-
3
197
KC820716.1
-
3
197
KC820717.1
-
3
197
KC820718.1
-
3
197
KC820719.1
-
3
197
KC820720.1
-
3
197
KC820721.1
-
3
197
KC820722.1
-
3
197
KC820723.1
-
3
197
KC820724.1
-
3
197
KC820725.1
-
3
197
KC820726.1
-
3
197
KC820727.1
-
3
197
KC820728.1
-
3
197
KC820729.1
-
3
197
KC820730.1
-
3
197
KC820732.1
-
3
197
KC820733.1
-
3
197
KC820734.1
-
3
197
KC820735.1
-
3
197
KC820736.1
-
3
197
KC820737.1
-
3
197
KC820739.1
-
3
197
KC820740.1
-
3
197
KC820741.1
-
3
197
KC820742.1
-
3
197
KC820743.1
-
3
197
KC820744.1
-
3
197
KC820745.1
-
3
197
KC820746.1
-
3
197
KC820747.1
-
3
197
KC820748.1
-
3
197
KC820749.1
-
3
197
KC820750.1
-
3
197
KC820751.1
-
3
197
KC820752.1
-
3
197
KC820753.1
-
3
197
KC820754.1
-
3
197
KC820755.1
-
3
197
KC820756.1
-
3
197
KC820757.1
-
3
197
KC820758.1
-
3
197
KC820759.1
-
2
86
KC820760.1
-
3
197
KC820761.1
-
3
197
KC820762.1
-
3
197
KC820763.1
-
3
197
KC820764.1
-
3
197
KC820765.1
-
3
197
KC820766.1
-
3
197
KC820767.1
-
3
197
KC820768.1
-
3
197
KC820769.1
-
3
197
KC820770.1
-
3
197
KC820771.1
-
3
197
KC820772.1
-
3
197
KC820773.1
-
3
197
KC820774.1
-
3
197
KC820775.1
-
3
197
KC820776.1
-
3
197
KC820777.1
-
3
197
KC820778.1
-
3
197
KC820779.1
-
3
197
KC820780.1
-
3
197
KC820781.1
-
3
197
KC820782.1
-
3
197
KC820783.1
-
3
197
KC820784.1
-
3
197
KC820786.1
-
3
197
M14694.1
-
7
827
M14695.1
-
7
823
X01405.1
-
4
403
X02469.1
-
7
837
X60010.1
-
6
701
X60011.1
-
6
702
X60012.1
-
6
702
X60013.1
-
6
702
X60015.1
-
6
702
X60017.1
-
6
702
X60018.1
-
6
702
X60019.1
-
6
702
X60020.1
-
6
702
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_003749209.
Panther Classification:
Molecular Function -
P53-like transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA strand renaturation
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
ER overload response
cell cycle arrest
Ras protein signal transduction
multicellular organism development
cell aging
protein localization
cell proliferation
negative regulation of cell proliferation
determination of adult lifespan
response to X-ray
response to gamma radiation
positive regulation of gene expression
viral process
protein sumoylation
cell differentiation
negative regulation of cell growth
DNA damage response, signal transduction by p53 class mediator
positive regulation of histone deacetylation
chromatin assembly
mitotic G1 DNA damage checkpoint
positive regulation of protein oligomerization
cellular response to UV
cellular response to drug
cellular response to glucose starvation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
entrainment of circadian clock by photoperiod
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of neuron apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
response to antibiotic
positive regulation of protein export from nucleus
regulation of mitochondrial membrane permeability
negative regulation of fibroblast proliferation
circadian behavior
positive regulation of peptidyl-tyrosine phosphorylation
negative regulation of helicase activity
protein tetramerization
negative regulation of telomerase activity
positive regulation of thymocyte apoptotic process
positive regulation of cell cycle arrest
cellular response to hypoxia
cellular response to ionizing radiation
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
replicative senescence
oxidative stress-induced premature senescence
intrinsic apoptotic signaling pathway
oligodendrocyte apoptotic process
positive regulation of execution phase of apoptosis
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
regulation of cell cycle G2/M phase transition
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
positive regulation of reactive oxygen species metabolic process
positive regulation of intrinsic apoptotic signaling pathway
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter sequence-specific DNA binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protease binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
copper ion binding
protein binding
ATP binding
transcription factor binding
zinc ion binding
enzyme binding
protein kinase binding
protein phosphatase binding
receptor tyrosine kinase binding
ubiquitin protein ligase binding
histone acetyltransferase binding
identical protein binding
sequence-specific DNA binding
protein self-association
transcription regulatory region DNA binding
protein heterodimerization activity
protein N-terminus binding
chaperone binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0