Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAF1 SIRNA Assays ›
          Gene Symbol:
          RAF1
          Gene Name:
          Raf-1 proto-oncogene, serine/threonine kinase
          Gene Aliases:
          CMD1NN, CRAF, NS5, Raf-1, c-Raf
          Chromosome Location:
          Chr.3: 12583601 - 12664201 on Build GRCh38
          UniGene ID:
          Hs.159130
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID VHS40468
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002880.3 NP_002871.1 13, 14 1820
          XM_005265355.2 XP_005265412.1 13, 14 1548
          XM_005265357.2 XP_005265414.1 12, 13 1637
          XM_005265358.4 XP_005265415.1 12, 13 1702
          XM_005265359.4 XP_005265416.1 11, 12 1603
          XM_011533974.2 XP_011532276.1 14, 15 1863
          XM_011533975.2 XP_011532277.1 13, 14 1752
          XM_017006966.1 XP_016862455.1 12, 13 1425
          GenBank mRNA AK303561.1 - 12, 13 1623
          AK303920.1 - 8, 9 897
          AK312248.1 - 13, 14 1714
          BC018119.2 - 13, 14 1734 3904404
          KU178297.1 - 12, 13 1405
          KU178298.1 - 13, 14 1465
          KU178299.1 - 11, 12 1306
          X03484.1 - 13, 14 1534

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          response to hypoxia
          stimulatory C-type lectin receptor signaling pathway
          protein phosphorylation
          apoptotic process
          signal transduction
          activation of adenylate cyclase activity
          heart development
          cell proliferation
          negative regulation of cell proliferation
          cell differentiation
          platelet activation
          thyroid gland development
          negative regulation of protein complex assembly
          positive regulation of peptidyl-serine phosphorylation
          ion transmembrane transport
          somatic stem cell population maintenance
          regulation of Rho protein signal transduction
          insulin secretion involved in cellular response to glucose stimulus
          response to muscle stretch
          wound healing
          regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          intermediate filament cytoskeleton organization
          regulation of cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          neurotrophin TRK receptor signaling pathway
          thymus development
          face development
          death-inducing signaling complex assembly
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          regulation of cell motility
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          protein binding
          ATP binding
          kinase activity
          small GTPase binding
          mitogen-activated protein kinase kinase binding
          identical protein binding
          metal ion binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0