Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATF2 SIRNA Assays ›
          Gene Symbol:
          ATF2
          Gene Name:
          activating transcription factor 2
          Gene Aliases:
          CRE-BP1, CREB-2, CREB2, HB16, TREB7
          Chromosome Location:
          Chr.2: 175072250 - 175168206 on Build GRCh38
          UniGene ID:
          Hs.592510
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID VHS40501
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256090.1 NP_001243019.1 9 869
          NM_001256091.1 NP_001243020.1 8 794
          NM_001256092.1 NP_001243021.1 7 737
          NM_001256093.1 NP_001243022.1 7 737
          NM_001256094.1 NP_001243023.1 8 834
          NM_001880.3 NP_001871.2 8 834
          NR_045768.1 9 869
          NR_045769.1 9 869
          NR_045770.1 9 869
          NR_045771.1 8 834
          NR_045772.1 8 772
          NR_045773.1 7 737
          NR_045774.1 8 834
          GenBank mRNA AK128731.1 - 8 734
          BC026175.1 - 8 801
          BC107698.1 - 7 704 6456836
          BC130335.1 - 7 600 40146366
          BC130337.1 - 7 600 40146368
          BX648469.1 - 7 624
          DQ003036.1 - 9 832
          DQ003037.1 - 8 797
          DQ003039.1 - 8 797
          DQ003040.1 - 8 735
          DQ003041.1 - 9 832
          DQ003042.1 - 8 735
          DQ003043.1 - 9 832
          DQ003044.1 - 9 832
          DQ003045.1 - 7 700
          DQ003046.1 - 7 700
          DQ003047.1 - 8 797
          DQ003048.1 - 7 700
          DQ003049.1 - 8 757
          X15875.1 - 6 561

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          outflow tract morphogenesis
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          response to osmotic stress
          cellular response to DNA damage stimulus
          response to water deprivation
          histone acetylation
          intra-S DNA damage checkpoint
          positive regulation of transforming growth factor beta2 production
          positive regulation of neuron apoptotic process
          fat cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of epithelial cell proliferation
          regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of sequence-specific DNA binding transcription factor activity
          adipose tissue development
          amelogenesis
          positive regulation of mitochondrial membrane permeability involved in apoptotic process
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcription factor activity, RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II activating transcription factor binding
          enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription coactivator activity
          histone acetyltransferase activity
          protein binding
          cAMP response element binding protein binding
          protein kinase binding
          cAMP response element binding
          metal ion binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0