Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARRB2 SIRNA Assays ›
          Gene Symbol:
          ARRB2
          Gene Name:
          arrestin beta 2
          Gene Aliases:
          ARB2, ARR2, BARR2
          Chromosome Location:
          Chr.17: 4710494 - 4721500 on Build GRCh38
          UniGene ID:
          Hs.435811
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID VHS40604
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARRB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257328.1 NP_001244257.1 14, 15 1434
          NM_001257329.1 NP_001244258.1 14, 15 1240
          NM_001257330.1 NP_001244259.1 14, 15 1407
          NM_001257331.1 NP_001244260.1 13, 14 1362
          NM_004313.3 NP_004304.1 14, 15 1371
          NM_199004.1 NP_945355.1 13, 14 1326
          NR_047516.1 13, 14 1340
          XM_006721520.1 XP_006721583.1 12, 13 1157
          XM_011523858.2 XP_011522160.1 14, 15 1256
          XM_017024645.1 XP_016880134.1 13, 14 1279
          GenBank mRNA AB209786.1 - 13, 14 1146
          AF106941.1 - 14, 15 1196
          AK097542.1 - 13, 14 1222
          AK290951.1 - 13, 14 1127
          AK297059.1 - 13, 14 1204
          AK297181.1 - 14, 15 1235
          BC007427.2 - 14, 15 1205 3028154
          BC039066.1 - 13, 14 1134
          BC067368.1 - 14, 15 1199 5748319
          BC095450.1 - 14, 15 1038
          CR450310.1 - 14, 15 1143
          CR749218.1 - 13, 14 1164
          DQ664180.1 - 14, 15 1179
          EU883572.1 - 14, 15 1143
          Z11501.1 - 14, 15 1143

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor internalization
          desensitization of G-protein coupled receptor protein signaling pathway by arrestin
          positive regulation of receptor internalization
          transcription from RNA polymerase II promoter
          transforming growth factor beta receptor signaling pathway
          dopamine receptor signaling pathway
          brain development
          adult walking behavior
          protein transport
          protein ubiquitination
          platelet activation
          negative regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          receptor internalization
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of synaptic transmission, dopaminergic
          negative regulation of interleukin-1 beta production
          negative regulation of interleukin-12 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of toll-like receptor signaling pathway
          negative regulation of GTPase activity
          negative regulation of smooth muscle cell apoptotic process
          follicle-stimulating hormone signaling pathway
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of natural killer cell mediated cytotoxicity
          positive regulation of peptidyl-tyrosine phosphorylation
          detection of temperature stimulus involved in sensory perception of pain
          positive regulation of protein kinase B signaling
          negative regulation of protein kinase B signaling
          positive regulation of calcium ion transport
          Wnt signaling pathway, planar cell polarity pathway
          cell chemotaxis
          regulation of androgen receptor signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of release of cytochrome c from mitochondria
          chemical synaptic transmission, postsynaptic
          positive regulation of DNA biosynthetic process
          G-protein coupled receptor binding
          receptor binding
          protein binding
          enzyme binding
          protein domain specific binding
          ubiquitin protein ligase binding
          alpha-1A adrenergic receptor binding
          alpha-1B adrenergic receptor binding
          angiotensin receptor binding
          type 1 angiotensin receptor binding
          D1 dopamine receptor binding
          follicle-stimulating hormone receptor binding
          type 2A serotonin receptor binding
          platelet activating factor receptor binding
          protein complex binding
          protein complex scaffold
          protein kinase B binding
          mitogen-activated protein kinase binding
          14-3-3 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0