Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEF2C SIRNA Assays ›
          Gene Symbol:
          MEF2C
          Gene Name:
          myocyte enhancer factor 2C
          Gene Aliases:
          C5DELq14.3, DEL5q14.3
          Chromosome Location:
          Chr.5: 88718241 - 88904105 on Build GRCh38
          UniGene ID:
          Hs.649965
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID VHS40929
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEF2C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001131005.2 NP_001124477.1 11 1521
          NM_001193347.1 NP_001180276.1 12 1581
          NM_001193348.1 NP_001180277.1 8 1093
          NM_001193350.1 NP_001180279.1 11 1773
          NM_001308002.1 NP_001294931.1 9 1237
          NM_002397.4 NP_002388.2 11 1776
          XM_005248511.2 XP_005248568.1 11 1310
          XM_006714619.2 XP_006714682.1 12 1632
          XM_006714625.3 XP_006714688.1 11 1639
          XM_011543396.2 XP_011541698.1 11 1361
          XM_011543397.2 XP_011541699.1 11 1542
          XM_017009475.1 XP_016864964.1 11 1603
          XM_017009476.1 XP_016864965.1 10 1585
          XM_017009477.1 XP_016864966.1 10 1524
          XM_017009478.1 XP_016864967.1 10 1579
          GenBank mRNA AK307883.1 - 10 1117
          AK312472.1 - 11 1481
          AL833268.1 - 12 1529
          AL833274.1 - 11 1470
          BC026341.1 - 11 1548
          FM163484.2 - 8 954
          L08895.1 - 11 1523
          S57212.1 - 1758

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_008760642, XM_006231731, XM_006231736, XM_006231733.
          This siRNA also targets the following transcripts in Mouse: NM_001170537, AK132573, L13171, BC057650, BC037731.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          blood vessel development
          osteoblast differentiation
          neuron migration
          B cell homeostasis
          heart looping
          endochondral ossification
          blood vessel remodeling
          chondrocyte differentiation
          germinal center formation
          regulation of germinal center formation
          primary heart field specification
          secondary heart field specification
          outflow tract morphogenesis
          sinoatrial valve morphogenesis
          cardiac ventricle formation
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          apoptotic process
          humoral immune response
          nervous system development
          heart development
          muscle organ development
          skeletal muscle tissue development
          muscle cell fate determination
          learning or memory
          response to virus
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of alkaline phosphatase activity
          neural crest cell differentiation
          cardiac muscle hypertrophy in response to stress
          myotube differentiation
          neuron differentiation
          platelet formation
          monocyte differentiation
          negative regulation of ossification
          melanocyte differentiation
          positive regulation of bone mineralization
          positive regulation of B cell proliferation
          cellular response to drug
          skeletal muscle cell differentiation
          cellular response to trichostatin A
          B cell proliferation
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          regulation of megakaryocyte differentiation
          positive regulation of myoblast differentiation
          positive regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of neurotransmitter secretion
          regulation of synaptic plasticity
          positive regulation of skeletal muscle tissue development
          neuron development
          cell morphogenesis involved in neuron differentiation
          embryonic viscerocranium morphogenesis
          negative regulation of epithelial cell proliferation
          B cell receptor signaling pathway
          smooth muscle cell differentiation
          positive regulation of muscle cell differentiation
          regulation of synapse assembly
          regulation of synaptic transmission, glutamatergic
          ventricular cardiac muscle cell differentiation
          palate development
          regulation of synaptic activity
          positive regulation of cardiac muscle cell proliferation
          excitatory postsynaptic potential
          regulation of sarcomere organization
          cartilage morphogenesis
          regulation of dendritic spine development
          renal tubule morphogenesis
          cellular response to lipopolysaccharide
          cellular response to calcium ion
          cellular response to parathyroid hormone stimulus
          cellular response to fluid shear stress
          cellular response to transforming growth factor beta stimulus
          positive regulation of cell proliferation in bone marrow
          glomerulus morphogenesis
          nephron tubule epithelial cell differentiation
          positive regulation of protein homodimerization activity
          positive regulation of macrophage apoptotic process
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          positive regulation of cardiac muscle cell differentiation
          positive regulation of behavioral fear response
          epithelial cell proliferation involved in renal tubule morphogenesis
          positive regulation of skeletal muscle cell differentiation
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          AT DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          activating transcription factor binding
          miRNA binding
          histone deacetylase binding
          transcription regulatory region DNA binding
          protein heterodimerization activity
          HMG box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0