Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
VHS40999
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
5
380
NM_001242560.1
NP_001229489.1
5
753
NM_004834.4
NP_004825.3
5
753
NM_145686.3
NP_663719.2
5
753
NM_145687.3
NP_663720.1
5
380
XM_005264048.2
XP_005264105.1
5
449
XM_005264050.3
XP_005264107.1
5
449
XM_005264052.3
XP_005264109.1
5
449
XM_005264057.3
XP_005264114.1
5
449
XM_005264058.3
XP_005264115.1
5
449
XM_005264059.3
XP_005264116.1
5
449
XM_005264060.3
XP_005264117.1
5
449
XM_005264062.3
XP_005264119.1
5
449
XM_005264063.3
XP_005264120.1
5
449
XM_005264064.3
XP_005264121.1
5
449
XM_005264065.3
XP_005264122.1
5
449
XM_005264066.3
XP_005264123.1
5
449
XM_005264068.3
XP_005264125.1
5
449
XM_005264069.2
XP_005264126.1
5
449
XM_005264071.2
XP_005264128.1
5
449
XM_006712865.2
XP_006712928.1
5
449
XM_006712868.2
XP_006712931.1
5
449
XM_006712869.2
XP_006712932.1
5
449
XM_017005349.1
XP_016860838.1
5
449
XM_017005350.1
XP_016860839.1
5
449
XM_017005351.1
XP_016860840.1
5
449
XM_017005352.1
XP_016860841.1
5
449
XM_017005353.1
XP_016860842.1
5
449
XM_017005354.1
XP_016860843.1
5
449
XM_017005355.1
XP_016860844.1
5
449
XM_017005356.1
XP_016860845.1
5
449
XM_017005357.1
XP_016860846.1
5
449
XM_017005358.1
XP_016860847.1
5
449
XM_017005359.1
XP_016860848.1
5
449
XM_017005360.1
XP_016860849.1
5
449
XM_017005361.1
XP_016860850.1
5
449
XM_017005362.1
XP_016860851.1
5
449
XM_017005363.1
XP_016860852.1
5
449
XM_017005364.1
XP_016860853.1
5
449
XM_017005365.1
XP_016860854.1
5
449
XM_017005366.1
XP_016860855.1
5
449
XM_017005367.1
XP_016860856.1
5
449
XM_017005368.1
XP_016860857.1
5
449
XM_017005369.1
XP_016860858.1
5
449
XM_017005370.1
XP_016860859.1
5
449
XM_017005371.1
XP_016860860.1
5
449
XM_017005372.1
XP_016860861.1
5
449
XM_017005373.1
XP_016860862.1
5
449
XM_017005374.1
XP_016860863.1
5
449
XM_017005375.1
XP_016860864.1
5
449
GenBank mRNA
AB014587.1
-
3
264
AF096300.1
-
5
454
AK295610.1
-
3
345
AK296396.1
-
5
449
AY212247.1
-
5
375
AY834276.1
-
5
375
EU794634.1
-
5
430
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline