Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other H19 SIRNA Assays ›
          Gene Symbol:
          H19
          Gene Name:
          H19, imprinted maternally expressed transcript (non-protein coding)
          Gene Aliases:
          ASM, ASM1, D11S813E
          Chromosome Location:
          Chr.1: 215744404 - 215747080 on Build Rnor_6.0
          UniGene ID:
          Rn.6171
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID n244246

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: H19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027324.1 1 808
          GenBank mRNA BC099104.1 - 1 729
          BC160921.1 - 1 542
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DY320105.1 - 48 Chr. - - [-] on Build
          GenBankmRNA FN804105.1 - 66 Chr. - - [-] on Build
          GenBankmRNA EV767378.1 - 113 Chr. - - [-] on Build
          GenBankmRNA FQ130424.1 - 71 Chr. - - [-] on Build
          GenBankmRNA DY318103.1 - 29 Chr. - - [-] on Build
          GenBankmRNA DY311238.1 - 30 Chr. - - [-] on Build
          GenBankmRNA EV765771.1 - 60 Chr. - - [-] on Build
          GenBankmRNA EV777243.1 - 50 Chr. - - [-] on Build
          GenBankmRNA DY310119.1 - 68 Chr. - - [-] on Build
          GenBankmRNA EV775529.1 - 71 Chr. - - [-] on Build
          GenBankmRNA EV780159.1 - 48 Chr. - - [-] on Build
          GenBankmRNA EV770822.1 - 31 Chr. - - [-] on Build
          GenBankmRNA DY310461.1 - 48 Chr. - - [-] on Build
          GenBankmRNA CV106090.1 - 42 Chr. - - [-] on Build
          GenBankmRNA CV117799.1 - 102 Chr. - - [-] on Build
          GenBankmRNA CV110176.1 - 71 Chr. - - [-] on Build
          GenBankmRNA EB464979.1 - 30 Chr. - - [-] on Build
          GenBankmRNA DY321092.1 - 43 Chr. - - [-] on Build
          GenBankmRNA CV118925.1 - 44 Chr. - - [-] on Build
          GenBankmRNA EV765989.1 - 76 Chr. - - [-] on Build
          GenBankmRNA EV774568.1 - 52 Chr. - - [-] on Build
          GenBankmRNA DY312033.1 - 45 Chr. - - [-] on Build
          GenBankmRNA DY309049.1 - 89 Chr. - - [-] on Build
          GenBankmRNA EV770556.1 - 122 Chr. - - [-] on Build
          GenBankmRNA EV780708.1 - 52 Chr. - - [-] on Build
          GenBankmRNA EV762440.1 - 62 Chr. - - [-] on Build
          GenBankmRNA EV763198.1 - 53 Chr. - - [-] on Build
          GenBankmRNA CV102461.1 - 125 Chr. - - [-] on Build

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          liver development
          skeletal muscle tissue development
          response to wounding
          response to nutrient levels
          response to lipid
          response to peptide hormone
          response to steroid hormone
          response to glucocorticoid
          mesenchymal to epithelial transition
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0