Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other H19 SIRNA Assays ›
Gene Symbol:
H19
Gene Name:
H19, imprinted maternally expressed transcript (non-protein coding)
Gene Aliases:
ASM, ASM1, D11S813E
Chromosome Location:
Chr.1: 215744404 - 215747080 on Build Rnor_6.0
UniGene ID:
Rn.6171
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
n244248
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: H19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027324.1
5
1754
GenBank mRNA
BC099104.1
-
5
1675
BC160921.1
-
5
1488
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
CB775863.1
-
202
Chr. - - [-] on Build
GenBankmRNA
DY321380.1
-
621
Chr. - - [-] on Build
GenBankmRNA
EV767862.1
-
327
Chr. - - [-] on Build
GenBankmRNA
EV779271.1
-
575
Chr. - - [-] on Build
GenBankmRNA
DY316134.1
-
619
Chr. - - [-] on Build
GenBankmRNA
DY309583.1
-
622
Chr. - - [-] on Build
GenBankmRNA
CB717003.1
-
333
Chr. - - [-] on Build
GenBankmRNA
CB777617.1
-
210
Chr. - - [-] on Build
GenBankmRNA
CB729441.1
-
190
Chr. - - [-] on Build
GenBankmRNA
CB727651.1
-
187
Chr. - - [-] on Build
GenBankmRNA
EB476291.1
-
642
Chr. - - [-] on Build
GenBankmRNA
CB694671.1
-
184
Chr. - - [-] on Build
GenBankmRNA
CB760807.1
-
222
Chr. - - [-] on Build
GenBankmRNA
FN804082.1
-
124
Chr. - - [-] on Build
GenBankmRNA
CB787770.1
-
359
Chr. - - [-] on Build
GenBankmRNA
EV777316.1
-
560
Chr. - - [-] on Build
GenBankmRNA
DY311398.1
-
619
Chr. - - [-] on Build
GenBankmRNA
CB734482.1
-
182
Chr. - - [-] on Build
GenBankmRNA
CB616745.1
-
396
Chr. - - [-] on Build
GenBankmRNA
FN804080.1
-
320
Chr. - - [-] on Build
GenBankmRNA
CB789056.1
-
312
Chr. - - [-] on Build
GenBankmRNA
CB811952.1
-
90
Chr. - - [-] on Build
GenBankmRNA
FN804109.1
-
680
Chr. - - [-] on Build
GenBankmRNA
CB778212.1
-
306
Chr. - - [-] on Build
GenBankmRNA
EV765723.1
-
614
Chr. - - [-] on Build
GenBankmRNA
DY308976.1
-
626
Chr. - - [-] on Build
GenBankmRNA
DY316528.1
-
609
Chr. - - [-] on Build
GenBankmRNA
CB795318.1
-
199
Chr. - - [-] on Build
GenBankmRNA
DY309786.1
-
626
Chr. - - [-] on Build
GenBankmRNA
AW918066.1
-
177
Chr. - - [-] on Build
GenBankmRNA
CB741497.1
-
317
Chr. - - [-] on Build
GenBankmRNA
CB768400.1
-
156
Chr. - - [-] on Build
GenBankmRNA
CB727024.1
-
200
Chr. - - [-] on Build
GenBankmRNA
EV779439.1
-
615
Chr. - - [-] on Build
GenBankmRNA
DY315928.1
-
556
Chr. - - [-] on Build
GenBankmRNA
BF554952.1
-
193
Chr. - - [-] on Build
GenBankmRNA
DY318779.1
-
626
Chr. - - [-] on Build
GenBankmRNA
DY319044.1
-
620
Chr. - - [-] on Build
GenBankmRNA
BF549021.1
-
193
Chr. - - [-] on Build
GenBankmRNA
CV104032.1
-
645
Chr. - - [-] on Build
GenBankmRNA
FN804083.1
-
341
Chr. - - [-] on Build
GenBankmRNA
DY309627.1
-
437
Chr. - - [-] on Build
GenBankmRNA
DY319382.1
-
76
Chr. - - [-] on Build
GenBankmRNA
CB727652.1
-
223
Chr. - - [-] on Build
GenBankmRNA
CV103161.1
-
433
Chr. - - [-] on Build
GenBankmRNA
FN804110.1
-
702
Chr. - - [-] on Build
GenBankmRNA
CV120053.1
-
609
Chr. - - [-] on Build
GenBankmRNA
CB737745.1
-
203
Chr. - - [-] on Build
GenBankmRNA
CB738492.1
-
137
Chr. - - [-] on Build
GenBankmRNA
FN804112.1
-
452
Chr. - - [-] on Build
GenBankmRNA
CB751384.1
-
423
Chr. - - [-] on Build
GenBankmRNA
FN804108.1
-
600
Chr. - - [-] on Build
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
liver development
skeletal muscle tissue development
response to wounding
response to nutrient levels
response to lipid
response to peptide hormone
response to steroid hormone
response to glucocorticoid
mesenchymal to epithelial transition
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0