Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other H19 SIRNA Assays ›
Gene Symbol:
H19
Gene Name:
H19, imprinted maternally expressed transcript (non-protein coding)
Gene Aliases:
ASM, ASM1, D11S813E
Chromosome Location:
Chr.1: 215744404 - 215747080 on Build Rnor_6.0
UniGene ID:
Rn.6171
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
n244253
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
404.00
Your Price
Online offer:
391.65
391.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: H19
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027324.1
3
1566
GenBank mRNA
BC099104.1
-
3
1487
BC160921.1
-
3
1300
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
CB775863.1
-
13
Chr. - - [-] on Build
GenBankmRNA
DY321380.1
-
433
Chr. - - [-] on Build
GenBankmRNA
EV767862.1
-
139
Chr. - - [-] on Build
GenBankmRNA
EV779271.1
-
387
Chr. - - [-] on Build
GenBankmRNA
DY316134.1
-
431
Chr. - - [-] on Build
GenBankmRNA
CB784671.1
-
436
Chr. - - [-] on Build
GenBankmRNA
EV768573.1
-
482
Chr. - - [-] on Build
GenBankmRNA
DY309583.1
-
434
Chr. - - [-] on Build
GenBankmRNA
CB717003.1
-
145
Chr. - - [-] on Build
GenBankmRNA
DY316240.1
-
580
Chr. - - [-] on Build
GenBankmRNA
DY319268.1
-
589
Chr. - - [-] on Build
GenBankmRNA
CB758044.1
-
252
Chr. - - [-] on Build
GenBankmRNA
EV762735.1
-
652
Chr. - - [-] on Build
GenBankmRNA
EB476291.1
-
454
Chr. - - [-] on Build
GenBankmRNA
EV764754.1
-
641
Chr. - - [-] on Build
GenBankmRNA
CV117164.1
-
613
Chr. - - [-] on Build
GenBankmRNA
EV768815.1
-
668
Chr. - - [-] on Build
GenBankmRNA
DY309618.1
-
438
Chr. - - [-] on Build
GenBankmRNA
EV764531.1
-
761
Chr. - - [-] on Build
GenBankmRNA
CV112487.1
-
645
Chr. - - [-] on Build
GenBankmRNA
CB787770.1
-
171
Chr. - - [-] on Build
GenBankmRNA
DY319936.1
-
451
Chr. - - [-] on Build
GenBankmRNA
DY311398.1
-
431
Chr. - - [-] on Build
GenBankmRNA
EV777316.1
-
372
Chr. - - [-] on Build
GenBankmRNA
DY309155.1
-
578
Chr. - - [-] on Build
GenBankmRNA
FN804111.1
-
445
Chr. - - [-] on Build
GenBankmRNA
DY317598.1
-
451
Chr. - - [-] on Build
GenBankmRNA
EV777202.1
-
645
Chr. - - [-] on Build
GenBankmRNA
CB616745.1
-
208
Chr. - - [-] on Build
GenBankmRNA
FN804080.1
-
132
Chr. - - [-] on Build
GenBankmRNA
CB707294.1
-
387
Chr. - - [-] on Build
GenBankmRNA
DY319344.1
-
583
Chr. - - [-] on Build
GenBankmRNA
CV105643.1
-
447
Chr. - - [-] on Build
GenBankmRNA
CB789056.1
-
124
Chr. - - [-] on Build
GenBankmRNA
FN804109.1
-
492
Chr. - - [-] on Build
GenBankmRNA
CB778212.1
-
118
Chr. - - [-] on Build
GenBankmRNA
EV770220.1
-
787
Chr. - - [-] on Build
GenBankmRNA
EV765723.1
-
426
Chr. - - [-] on Build
GenBankmRNA
CA510644.1
-
614
Chr. - - [-] on Build
GenBankmRNA
CB745847.1
-
281
Chr. - - [-] on Build
GenBankmRNA
DY308976.1
-
438
Chr. - - [-] on Build
GenBankmRNA
CB788342.1
-
213
Chr. - - [-] on Build
GenBankmRNA
DY308870.1
-
392
Chr. - - [-] on Build
GenBankmRNA
FQ117437.1
-
716
Chr. - - [-] on Build
GenBankmRNA
FN804104.1
-
681
Chr. - - [-] on Build
GenBankmRNA
EV763246.1
-
464
Chr. - - [-] on Build
GenBankmRNA
CV111815.1
-
579
Chr. - - [-] on Build
GenBankmRNA
CV108855.1
-
437
Chr. - - [-] on Build
GenBankmRNA
CB776874.1
-
267
Chr. - - [-] on Build
GenBankmRNA
EV770283.1
-
489
Chr. - - [-] on Build
GenBankmRNA
DY316528.1
-
421
Chr. - - [-] on Build
GenBankmRNA
DY309786.1
-
438
Chr. - - [-] on Build
GenBankmRNA
DY319332.1
-
659
Chr. - - [-] on Build
GenBankmRNA
EV765620.1
-
731
Chr. - - [-] on Build
GenBankmRNA
CB741497.1
-
130
Chr. - - [-] on Build
GenBankmRNA
CB802494.1
-
354
Chr. - - [-] on Build
GenBankmRNA
CB798837.1
-
409
Chr. - - [-] on Build
GenBankmRNA
EV779439.1
-
427
Chr. - - [-] on Build
GenBankmRNA
DY315928.1
-
368
Chr. - - [-] on Build
GenBankmRNA
CB737744.1
-
303
Chr. - - [-] on Build
GenBankmRNA
FQ116831.1
-
653
Chr. - - [-] on Build
GenBankmRNA
DY319044.1
-
432
Chr. - - [-] on Build
GenBankmRNA
DY318779.1
-
438
Chr. - - [-] on Build
GenBankmRNA
CB741500.1
-
380
Chr. - - [-] on Build
GenBankmRNA
CV104032.1
-
457
Chr. - - [-] on Build
GenBankmRNA
FN804083.1
-
153
Chr. - - [-] on Build
GenBankmRNA
EV767032.1
-
682
Chr. - - [-] on Build
GenBankmRNA
DY318792.1
-
604
Chr. - - [-] on Build
GenBankmRNA
EV775200.1
-
639
Chr. - - [-] on Build
GenBankmRNA
DY309627.1
-
249
Chr. - - [-] on Build
GenBankmRNA
EV774568.1
-
810
Chr. - - [-] on Build
GenBankmRNA
DY313825.1
-
580
Chr. - - [-] on Build
GenBankmRNA
FM029648.1
-
438
Chr. - - [-] on Build
GenBankmRNA
CB729442.1
-
375
Chr. - - [-] on Build
GenBankmRNA
FN804110.1
-
514
Chr. - - [-] on Build
GenBankmRNA
CV103161.1
-
245
Chr. - - [-] on Build
GenBankmRNA
CV120053.1
-
421
Chr. - - [-] on Build
GenBankmRNA
CB786289.1
-
390
Chr. - - [-] on Build
GenBankmRNA
EV768581.1
-
657
Chr. - - [-] on Build
GenBankmRNA
CV115681.1
-
579
Chr. - - [-] on Build
GenBankmRNA
CB732670.1
-
312
Chr. - - [-] on Build
GenBankmRNA
FN804112.1
-
264
Chr. - - [-] on Build
GenBankmRNA
CB751384.1
-
235
Chr. - - [-] on Build
GenBankmRNA
CV105546.1
-
519
Chr. - - [-] on Build
GenBankmRNA
CB758045.1
-
382
Chr. - - [-] on Build
GenBankmRNA
CV113239.1
-
654
Chr. - - [-] on Build
GenBankmRNA
EV769573.1
-
679
Chr. - - [-] on Build
GenBankmRNA
EV762440.1
-
818
Chr. - - [-] on Build
GenBankmRNA
CB741501.1
-
296
Chr. - - [-] on Build
GenBankmRNA
FN804108.1
-
412
Chr. - - [-] on Build
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
liver development
skeletal muscle tissue development
response to wounding
response to nutrient levels
response to lipid
response to peptide hormone
response to steroid hormone
response to glucocorticoid
mesenchymal to epithelial transition
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0