Search
Search

BC029722| Gene Symbol: BC029722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015528.1 | 2 | 292 | |||
| GenBank mRNA | AK009222.1 | - | 2 | 330 | ||
| AK157030.1 | - | 2 | 276 | |||
| AK158030.1 | - | 2 | 293 | |||
| AK159419.1 | - | 2 | 325 | |||
| BC029722.1 | - | 2 | 292 | |||