Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SVIP SIRNA Assays ›
          Gene Symbol:
          Svip
          Gene Name:
          small VCP/p97-interacting protein
          Gene Aliases:
          1700006C13Rik, 6620401M08Rik
          Chromosome Location:
          Chr.7: 51997161 - 52006018 on Build GRCm38
          UniGene ID:
          Mm.386823
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n254870

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Svip
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027704.1 1 111
          GenBank mRNA AK005663.1 - 1 89
          AK029831.1 - 1 89
          BC049720.1 - 95
          BC100498.1 - 94
          BY089978.1 - 1 111
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CA463181.1 - 1 83 Chr. 7 - 51999577 - 52005686 [-] on Build GRCm38
          GenBankmRNA BU946028.1 - 1 91 Chr. 7 - 51999577 - 52005691 [-] on Build GRCm38
          GenBankmRNA BU961998.1 - 98 Chr. - - [-] on Build
          GenBankmRNA BU938089.1 - 86 Chr. - - [-] on Build
          GenBankmRNA CN835687.1 - 1 91 Chr. 7 - 51999610 - 52005691 [-] on Build GRCm38
          GenBankmRNA BY705852.1 - 1 86 Chr. 7 - 51999610 - 52005689 [-] on Build GRCm38
          GenBankmRNA BU936203.1 - 86 Chr. - - [-] on Build
          GenBankmRNA CA464693.1 - 1 84 Chr. 7 - 51999608 - 52005686 [-] on Build GRCm38
          GenBankmRNA CN842957.1 - 101 Chr. - - [-] on Build
          GenBankmRNA CA464522.1 - 1 91 Chr. 7 - 51999610 - 52005691 [-] on Build GRCm38
          GenBankmRNA CN831979.1 - 89 Chr. - - [-] on Build
          GenBankmRNA BU936457.1 - 108 Chr. - - [-] on Build
          GenBankmRNA BU937560.1 - 1 91 Chr. 7 - 51999581 - 52005691 [-] on Build GRCm38
          GenBankmRNA CA465491.1 - 1 91 Chr. 7 - 51999594 - 52005691 [-] on Build GRCm38
          GenBankmRNA BU937269.1 - 91 Chr. - - [-] on Build
          GenBankmRNA BU936830.1 - 1 84 Chr. 7 - 51999608 - 52005686 [-] on Build GRCm38
          GenBankmRNA CN838154.1 - 91 Chr. - - [-] on Build
          GenBankmRNA BY716085.1 - 1 86 Chr. 7 - 51999730 - 52005689 [-] on Build GRCm38
          GenBankmRNA CN837221.1 - 1 84 Chr. 7 - 51999608 - 52005686 [-] on Build GRCm38
          GenBankmRNA CN843771.1 - 1 97 Chr. 7 - 51999606 - 52005700 [-] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of autophagy
          negative regulation of protein complex assembly
          positive regulation of protein lipidation
          negative regulation of ER-associated ubiquitin-dependent protein catabolic process
          negative regulation of retrograde protein transport, ER to cytosol
          protein self-association

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0