Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RN5S SIRNA Assays ›
          Gene Symbol:
          Rn5s Gm22109 Gm22291 Gm23284 Gm25018 Gm25212 Gm26391 Tmem38b n-R5s100 n-R5s103 n-R5s104 n-R5s105 n-R5s108 n-R5s110 n-R5s111 n-R5s113 n-R5s117 n-R5s121 n-R5s122 n-R5s123 n-R5s124 n-R5s128 n-R5s133 n-R5s134 n-R5s136 n-R5s138 n-R5s139 n-R5s141 n-R5s142 n-R5s143 n-R5s144 n-R5s146 n-R5s149
          Gene Name:
          5S RNA
          predicted gene, 22109
          predicted gene, 22291
          predicted gene, 23284
          predicted gene, 25018
          predicted gene, 25212
          predicted gene, 26391
          transmembrane protein 38B
          nuclear encoded rRNA 5S 100
          nuclear encoded rRNA 5S 103
          nuclear encoded rRNA 5S 104
          nuclear encoded rRNA 5S 105
          nuclear encoded rRNA 5S 108
          nuclear encoded rRNA 5S 110
          nuclear encoded rRNA 5S 111
          nuclear encoded rRNA 5S 113
          nuclear encoded rRNA 5S 117
          nuclear encoded rRNA 5S 121
          nuclear encoded rRNA 5S 122
          nuclear encoded rRNA 5S 123
          nuclear encoded rRNA 5S 124
          nuclear encoded rRNA 5S 128
          nuclear encoded rRNA 5S 133
          nuclear encoded rRNA 5S 134
          nuclear encoded rRNA 5S 136
          nuclear encoded rRNA 5S 138
          nuclear encoded rRNA 5S 139
          nuclear encoded rRNA 5S 141
          nuclear encoded rRNA 5S 142
          nuclear encoded rRNA 5S 143
          nuclear encoded rRNA 5S 144
          nuclear encoded rRNA 5S 146
          nuclear encoded rRNA 5S 149
          Gene Aliases:
          -
          Rn5s23
          Rn5s30
          Rn5s31
          Rn5s10
          Rn5s28
          Rn5s17
          1600017F22Rik, AA437809, AV051057, D4Ertd89e, TRIC-B, mg33b
          Rn5s2
          Rn5s3
          Rn5s4
          Rn5s5
          Rn5s6
          Rn5s7
          Rn5s8
          Rn5s9
          Rn5s11
          Rn5s12
          Rn5s13
          Rn5s14
          Rn5s15
          Rn5s16
          Rn5s18
          Rn5s19
          Rn5s20
          Rn5s21
          Rn5s22
          Rn5s24
          Rn5s25
          Rn5s26
          Rn5s27
          Rn5s29
          Rn5s32
          Chromosome Location:
          Chr.8: 123598156 - 123598276 on Build GRCm38
          Chromosome Location:
          Chr.8: 123606611 - 123606731 on Build GRCm38
          Chromosome Location:
          Chr.8: 123618534 - 123618654 on Build GRCm38
          Chromosome Location:
          Chr.8: 123620239 - 123620359 on Build GRCm38
          Chromosome Location:
          Chr.8: 123565967 - 123566087 on Build GRCm38
          Chromosome Location:
          Chr.8: 123615097 - 123615217 on Build GRCm38
          Chromosome Location:
          Chr.8: 123593069 - 123593189 on Build GRCm38
          Chromosome Location:
          Chr.4: 53826045 - 53862019 on Build GRCm38
          Chromosome Location:
          Chr.8: 123538784 - 123538904 on Build GRCm38
          Chromosome Location:
          Chr.8: 123543871 - 123543991 on Build GRCm38
          Chromosome Location:
          Chr.8: 123545576 - 123545696 on Build GRCm38
          Chromosome Location:
          Chr.8: 123547263 - 123547383 on Build GRCm38
          Chromosome Location:
          Chr.8: 123552362 - 123552482 on Build GRCm38
          Chromosome Location:
          Chr.8: 123555761 - 123555881 on Build GRCm38
          Chromosome Location:
          Chr.8: 123557488 - 123557608 on Build GRCm38
          Chromosome Location:
          Chr.8: 123560886 - 123561006 on Build GRCm38
          Chromosome Location:
          Chr.8: 123567635 - 123567755 on Build GRCm38
          Chromosome Location:
          Chr.8: 123574395 - 123574515 on Build GRCm38
          Chromosome Location:
          Chr.8: 123576074 - 123576194 on Build GRCm38
          Chromosome Location:
          Chr.8: 123577753 - 123577873 on Build GRCm38
          Chromosome Location:
          Chr.8: 123579448 - 123579568 on Build GRCm38
          Chromosome Location:
          Chr.8: 123586240 - 123586360 on Build GRCm38
          Chromosome Location:
          Chr.8: 123594752 - 123594872 on Build GRCm38
          Chromosome Location:
          Chr.8: 123596455 - 123596575 on Build GRCm38
          Chromosome Location:
          Chr.8: 123599859 - 123599979 on Build GRCm38
          Chromosome Location:
          Chr.8: 123603245 - 123603365 on Build GRCm38
          Chromosome Location:
          Chr.8: 123604946 - 123605066 on Build GRCm38
          Chromosome Location:
          Chr.8: 123608318 - 123608438 on Build GRCm38
          Chromosome Location:
          Chr.8: 123610015 - 123610135 on Build GRCm38
          Chromosome Location:
          Chr.8: 123611713 - 123611833 on Build GRCm38
          Chromosome Location:
          Chr.8: 123613412 - 123613532 on Build GRCm38
          Chromosome Location:
          Chr.8: 123616808 - 123616928 on Build GRCm38
          Chromosome Location:
          Chr.8: 123621926 - 123622046 on Build GRCm38
          UniGene ID:
          -
          -
          -
          -
          -
          -
          -
          Mm.27184
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID n256117

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rn5s
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_030686.1 72
          Gene Symbol: Gm22109
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046147.1 72
          Gene Symbol: Gm22291
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046154.1 72
          Gene Symbol: Gm23284
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046155.1 72
          Gene Symbol: Gm25018
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046127.1 72
          Gene Symbol: Gm25212
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046152.1 72
          Gene Symbol: Gm26391
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046140.1 72
          Gene Symbol: Tmem38b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK136131.1 - 5 1316
          AK150377.1 - 6 1544
          Gene Symbol: n-R5s100
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046118.1 72
          Gene Symbol: n-R5s103
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046119.1 72
          Gene Symbol: n-R5s104
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046120.1 72
          Gene Symbol: n-R5s105
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046122.1 72
          Gene Symbol: n-R5s108
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046123.1 72
          Gene Symbol: n-R5s110
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046124.1 72
          Gene Symbol: n-R5s111
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046125.1 72
          Gene Symbol: n-R5s113
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046126.1 72
          Gene Symbol: n-R5s117
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046128.1 72
          Gene Symbol: n-R5s121
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046129.1 72
          Gene Symbol: n-R5s122
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046130.1 72
          Gene Symbol: n-R5s123
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046132.1 72
          Gene Symbol: n-R5s124
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046133.1 72
          Gene Symbol: n-R5s128
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046134.1 72
          Gene Symbol: n-R5s133
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046141.1 72
          Gene Symbol: n-R5s134
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046143.1 72
          Gene Symbol: n-R5s136
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046144.1 1 72
          Gene Symbol: n-R5s138
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046145.1 72
          Gene Symbol: n-R5s139
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046146.1 72
          Gene Symbol: n-R5s141
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046148.1 72
          Gene Symbol: n-R5s142
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046149.1 72
          Gene Symbol: n-R5s143
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046150.1 72
          Gene Symbol: n-R5s144
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046151.1 72
          Gene Symbol: n-R5s146
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046153.1 72
          Gene Symbol: n-R5s149
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046156.1 1 72

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NR_023371, NR_023369, NR_023372, NR_023368, NR_023374, and others.
          This siRNA also targets the following transcripts in Rat: FQ217280, FQ227942, FQ212247.

          Panther Classification:

          Molecular Function -

          ion channel

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-czsl9:80/100.66.77.209:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0