Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033282.1 | - | 17 | 3265 | ||
AK040460.1 | - | 15 | 2943 | |||
AK052234.1 | - | 17 | 3309 | |||
AK054332.1 | - | 17 | 3310 | |||
AK083413.1 | - | 17 | 3308 | |||
AK084546.1 | - | 17 | 3307 | |||
AK084604.1 | - | 17 | 3307 | |||
AK085903.1 | - | 15 | 2972 | |||
AK088044.1 | - | 17 | 4057 | |||
AK129006.1 | - | 17 | 4054 | |||
AK159694.1 | - | 19 | 4062 | |||
AK168736.1 | - | 16 | 3860 | |||
AK192828.1 | - | 1 | 193 | |||
BC008518.1 | - | 17 | 4040 | 3601702 |